ADVH49B81F55 AChemBlock

CHIR-090

  • Cas Number : 728865-23-4
  • Molecular Weight : 437.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/chir-090.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72357
Availability details for
Quantity
  • Availability
  • Add to Cart