ADVH49B81F57 AChemBlock

4-(bromomethyl)-2,2-difluoro-2H-1,3-benzodioxole

  • Cas Number : 72769-05-2
  • Molecular Weight : 251.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromomethyl-2-2-difluoro-2h-1-3-benzodioxole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74358
Availability details for
Quantity
  • Availability
  • Add to Cart