ADVH49B81F58 AChemBlock

2-chloro-3-fluoro-5-(trifluoromethyl)pyridine

  • Cas Number : 72600-67-0
  • Molecular Weight : 199.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-3-fluoro-5-trifluoromethyl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74366
Availability details for
Quantity
  • Availability
  • Add to Cart