ADVH49B81F5C AChemBlock

quinoline-5-carboxylic acid

  • Cas Number : 7250-53-5
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/quinoline-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71111
Availability details for
Quantity
  • Availability
  • Add to Cart