ADVH49B81F5E AChemBlock

3-fluoro-5-iodobenzonitrile

  • Cas Number : 723294-75-5
  • Molecular Weight : 247.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-fluoro-5-iodobenzonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74378
Availability details for
Quantity
  • Availability
  • Add to Cart