ADVH49B81F61 AChemBlock

NSC319726

  • Cas Number : 71555-25-4
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/nsc319726.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86748
Availability details for
Quantity
  • Availability
  • Add to Cart