ADVH49B81F6A AChemBlock

2-Hydroxy-6-methoxybenzaldehyde

  • Cas Number : 700-44-7
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-hydroxy-6-methoxybenzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86496
Availability details for
Quantity
  • Availability
  • Add to Cart