ADVH49B81F6D AChemBlock

5-methyl-1,2,3,4-tetrahydronaphthalen-1-one

  • Cas Number : 6939-35-1
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-1-2-3-4-tetrahydronaphthalen-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74496
Availability details for
Quantity
  • Availability
  • Add to Cart