ADVH49B81F71 AChemBlock

3,5-Diamino-1H-pyrazole-4-carbonitrile

  • Cas Number : 6844-58-2
  • Molecular Weight : 123.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.achemblock.com/3-5-diamino-1h-pyrazole-4-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72324
Availability details for
Quantity
  • Availability
  • Add to Cart