ADVH49B81F72 AChemBlock

Afimoxifene

  • Cas Number : 68392-35-8
  • Molecular Weight : 387.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/afimoxifene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86608
Availability details for
Quantity
  • Availability
  • Add to Cart