ADVH49B81F74 AChemBlock

5-(difluoromethyl)-1H-pyrazole-3-carboxylic acid

  • Cas Number : 681034-51-5
  • Molecular Weight : 162.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-difluoromethyl-1h-pyrazole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74549
Availability details for
Quantity
  • Availability
  • Add to Cart