ADVH49B81F75 AChemBlock

4-(2-aminoethyl)-3-[(3R)-6,8-difluorochroman-3-yl]-1H-imidazole-2-thione hydrochloride

  • Cas Number : 677773-32-9
  • Molecular Weight : 347.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-2-aminoethyl-3-3r-6-8-difluorochroman-3-yl-1h-imidazole-2-thione-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74559
Availability details for
Quantity
  • Availability
  • Add to Cart