ADVH49B81F7B AChemBlock

3-Acetyl-6-azaindole

  • Cas Number : 67058-71-3
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-acetyl-6-azaindole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72411
Availability details for
Quantity
  • Availability
  • Add to Cart