ADVH49B81F7E AChemBlock

4-[(6-chloro-3-pyridyl)oxy]-6,7-dimethoxy-quinoline

  • Cas Number : 666731-22-2
  • Molecular Weight : 316.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-6-chloro-3-pyridyl-oxy-6-7-dimethoxy-quinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74601
Availability details for
Quantity
  • Availability
  • Add to Cart