ADVH49B81F80 AChemBlock

(4R)-3-[(tert-butoxy)carbonyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid

  • Cas Number : 660852-86-8
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4r-3-tert-butoxy-carbonyl-2-2-dimethyl-1-3-oxazolidine-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74627
Availability details for
Quantity
  • Availability
  • Add to Cart