ADVH49B81F81 AChemBlock

2-bromo-3,5-difluoropyridine

  • Cas Number : 660425-16-1
  • Molecular Weight : 193.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-3-5-difluoropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71146
Availability details for
Quantity
  • Availability
  • Add to Cart