ADVH49B81F82 AChemBlock

3-Methyl-1,2-benzisoxazol-6-ol

  • Cas Number : 66033-92-9
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1-2-benzisoxazol-6-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86497
Availability details for
Quantity
  • Availability
  • Add to Cart