ADVH49B81F84 AChemBlock

AMG-517

  • Cas Number : 659730-32-2
  • Molecular Weight : 430.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/amg-517.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86733
Availability details for
Quantity
  • Availability
  • Add to Cart