ADVH49B81F85 AChemBlock

6-phenoxybenzo[d]thiazol-2-amine

  • Cas Number : 65948-19-8
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-phenoxybenzo-d-thiazol-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86548
Availability details for
Quantity
  • Availability
  • Add to Cart