ADVH49B81F87 AChemBlock

2,6-dihydro-1,2-benzoxazol-6-one

  • Cas Number : 65685-55-4
  • Molecular Weight : 135.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-6-dihydro-1-2-benzoxazol-6-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74641
Availability details for
Quantity
  • Availability
  • Add to Cart