ADVH49B81F8A AChemBlock

ethyl 4-chloro-2-methoxypyrimidine-5-carboxylate

  • Cas Number : 65269-18-3
  • Molecular Weight : 216.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-chloro-2-methoxypyrimidine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86507
Availability details for
Quantity
  • Availability
  • Add to Cart