ADVH49B81F8F AChemBlock

1-bromo-5-fluoro-2-methyl-4-nitrobenzene

  • Cas Number : 64695-96-1
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-bromo-5-fluoro-2-methyl-4-nitrobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71157
Availability details for
Quantity
  • Availability
  • Add to Cart