ADVH49B81F92 AChemBlock

3,4-dichloro-5-methoxy-pyridazine

  • Cas Number : 63910-32-7
  • Molecular Weight : 179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-4-dichloro-5-methoxy-pyridazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74702
Availability details for
Quantity
  • Availability
  • Add to Cart