ADVH49B81F95 AChemBlock

methyl 5-methoxy-2-methyl-pyridine-3-carboxylate

  • Cas Number : 63820-73-5
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-methoxy-2-methyl-pyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74709
Availability details for
Quantity
  • Availability
  • Add to Cart