ADVH49B81F97 AChemBlock

1,4-dibromo-2,5-diiodobenzene

  • Cas Number : 63262-06-6
  • Molecular Weight : 487.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-dibromo-2-5-diiodobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71168
Availability details for
Quantity
  • Availability
  • Add to Cart