ADVH49B81F99 AChemBlock

Ethyl 4-pentynoate

  • Cas Number : 63093-41-4
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-pentynoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86253
Availability details for
Quantity
  • Availability
  • Add to Cart