ADVH49B81F9E AChemBlock

2-(chloromethyl)-4-methoxypyridine hydrochloride

  • Cas Number : 62734-08-1
  • Molecular Weight : 194.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-chloromethyl-4-methoxypyridine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74750
Availability details for
Quantity
  • Availability
  • Add to Cart