ADVH49B81FA0 AChemBlock

rhodamine 123

  • Cas Number : 62669-70-9
  • Molecular Weight : 380.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/rhodamine-123.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86591
Availability details for
Quantity
  • Availability
  • Add to Cart