ADVH49B81FA7 AChemBlock

4-Amino-benzenepropanamine dihydrochloride

  • Cas Number : 61798-01-4
  • Molecular Weight : 223.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-amino-benzenepropanamine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70323
Availability details for
Quantity
  • Availability
  • Add to Cart