ADVH49B81FB0 AChemBlock

4-(4,5-Dihydro-1H-imidazol-2-yl)-phenylamine

  • Cas Number : 61033-71-4
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-5-dihydro-1h-imidazol-2-yl-phenylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86734
Availability details for
Quantity
  • Availability
  • Add to Cart