ADVH49B81FB3 AChemBlock

5-chloro-1-methyl-2,3-dihydro-1H-indole-2,3-dione

  • Cas Number : 60434-13-1
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-1-methyl-2-3-dihydro-1h-indole-2-3-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74855
Availability details for
Quantity
  • Availability
  • Add to Cart