ADVH49B81FB7 AChemBlock

3-methyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride

  • Cas Number : 601515-50-8
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-5-6-7-8-tetrahydroimidazo-1-5-a-pyrazine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74871
Availability details for
Quantity
  • Availability
  • Add to Cart