ADVH49B81FBA AChemBlock

3-Methyl-N-(hydroxymethyl)benzamide

  • Cas Number : 59630-86-3
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-n-hydroxymethyl-benzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86696
Availability details for
Quantity
  • Availability
  • Add to Cart