ADVH49B81FBB AChemBlock

methyl 6-chloro-5-nitropyridine-3-carboxylate

  • Cas Number : 59237-53-5
  • Molecular Weight : 216.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-chloro-5-nitropyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71207
Availability details for
Quantity
  • Availability
  • Add to Cart