ADVH49B81FD0 AChemBlock

7-hydroxy-4H,5H-pyrazolo[1,5-a]pyrimidin-5-one

  • Cas Number : 57489-70-0
  • Molecular Weight : 151.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-hydroxy-4h-5h-pyrazolo-1-5-a-pyrimidin-5-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74986
Availability details for
Quantity
  • Availability
  • Add to Cart