ADVH49B81FD5 AChemBlock

4-Chlorophenethyl iodide

  • Cas Number : 57186-64-8
  • Molecular Weight : 266.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-chlorophenethyl-iodide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70328
Availability details for
Quantity
  • Availability
  • Add to Cart