ADVH49B81FD6 AChemBlock

1-(3-methoxyphenyl)guanidine

  • Cas Number : 57004-60-1
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-methoxyphenyl-guanidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86513
Availability details for
Quantity
  • Availability
  • Add to Cart