ADVH49B81FD8 AChemBlock

2-(4-chlorophenyl)imidazo[1,2-a]pyrimidine

  • Cas Number : 56921-86-9
  • Molecular Weight : 229.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-4-chlorophenyl-imidazo-1-2-a-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75009
Availability details for
Quantity
  • Availability
  • Add to Cart