ADVH49B81FDE AChemBlock

2-methylindolin-7-amine

  • Cas Number : 55899-44-0
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methylindolin-7-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75059
Availability details for
Quantity
  • Availability
  • Add to Cart