ADVH49B81FDF AChemBlock

3,4-dihydro-2H-1-benzopyran-6-carbaldehyde

  • Cas Number : 55745-97-6
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-4-dihydro-2h-1-benzopyran-6-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75070
Availability details for
Quantity
  • Availability
  • Add to Cart