ADVH49B81FE1 AChemBlock

3-Formyl-4-hydroxybenzoic acid benzyl ester

  • Cas Number : 550346-26-4
  • Molecular Weight : 256.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-formyl-4-hydroxybenzoic-acid-benzyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86790
Availability details for
Quantity
  • Availability
  • Add to Cart