ADVH49B81FE2 AChemBlock

4,5,6,7-tetrahydrothieno[3,2-c]pyridine

  • Cas Number : 54903-50-3
  • Molecular Weight : 139.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-5-6-7-tetrahydrothieno-3-2-c-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71242
Availability details for
Quantity
  • Availability
  • Add to Cart