ADVH49B81FE7 AChemBlock

(8-bromooctyl)benzene

  • Cas Number : 54646-75-2
  • Molecular Weight : 269.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-bromooctyl-benzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86576
Availability details for
Quantity
  • Availability
  • Add to Cart