ADVH49B81FEC AChemBlock

1,3,3-trimethylindolin-6-amine

  • Cas Number : 545393-67-7
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-3-3-trimethylindolin-6-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75133
Availability details for
Quantity
  • Availability
  • Add to Cart