ADVH49B81FED AChemBlock

1-ethylsulfonyl-4-(4-nitrophenyl)piperazine

  • Cas Number : 544462-63-7
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-ethylsulfonyl-4-4-nitrophenyl-piperazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75136
Availability details for
Quantity
  • Availability
  • Add to Cart