ADVH49B81FEF AChemBlock

6-bromo-2-methylquinazolin-4(1H)-one

  • Cas Number : 5426-59-5
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-methylquinazolin-4-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86529
Availability details for
Quantity
  • Availability
  • Add to Cart