ADVH49B81FF3 AChemBlock

1-(p-tolyl)guanidine

  • Cas Number : 54015-04-2
  • Molecular Weight : 149.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-p-tolyl-guanidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86522
Availability details for
Quantity
  • Availability
  • Add to Cart