ADVH49B81FF7 AChemBlock

[(1H-indol-1-yl)methyl]dimethylamine

  • Cas Number : 5379-79-3
  • Molecular Weight : 174.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1h-indol-1-yl-methyl-dimethylamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75173
Availability details for
Quantity
  • Availability
  • Add to Cart