ADVH49B81FF9 AChemBlock

6-chloro-1H-indole-2-carbaldehyde

  • Cas Number : 53590-59-3
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1h-indole-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86549
Availability details for
Quantity
  • Availability
  • Add to Cart