ADVH49B8200F AChemBlock

4-hydrazinyl-1-methylpiperidine

  • Cas Number : 51304-64-4
  • Molecular Weight : 129.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-hydrazinyl-1-methylpiperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75294
Availability details for
Quantity
  • Availability
  • Add to Cart